Learn More
Bioland Scientific LLC Endofree plasmid miniprep II kit (50 preps)
SDP
Supplier:  Bioland Scientific LLC PD0211
Encompass
This kit is designed for fast and efficient purification of plasmid DNA from 3 to 12 mL of E. coli culture. The mini column II has a DNA binding capacity of 80 µg. The purified endofree DNA is ready for downstream applications such as transfection of endotoxin-sensitive cell lines, primary cultured cells or microinjection.
Catalog No. NC3781122