Learn More
ILEX LIFE SCIENCES LLC LIONEX EndoTrap Empty Columns Small (for 0.2-2 mL resin), pack of 5, endotoxin-removal accessory
SDP
Supplier:  ILEX LIFE SCIENCES LLC LET0021
Encompass_Preferred
EndoGrade Endotoxin-Free Glass Test Tubes (112 ct.), manufactured in Germany by LIONEX GmbH. 112 endotoxin-free borosilicate glass test tubes with aluminum screw cap. CAPACITY: 5 ml. HEIGHT: 50 mm. APPLICATIONS: EndoGrade test tubes are useful to dilute or aliquot samples without introducing LPS contamination. EndoGrade test tubes are ideal for diluting endotoxin standards when performing endotoxin testing (e.g. EndoLISA, EndoZyme). EndoGrade test tubes may also be used to store recovered samples after endotoxin removal with EndoTrap or other LPS-removal systems.
Catalog No. 50-233-9587