Learn More
World Precision Instrument Cell Culture Cup Chambers for TEER Measurement-ENDOHM-12G
SDP
Supplier:  World Precision Instrument ENDOHM12G
Encompass
These Cell Culture Cup Chambers used in TEER measurement for removable culture cup systems using EVOM2 meters for endothelial and epithelial cell cultures.
NOTE: These EndOhms are for use with older EVOM™ meters. For EndOhms used with the new EVOM™ Manual (or EVOM3), see EVOM™ Electrodes for TEER.
EndOhm chambers provide reproducible resistance measurements of endothelial and epithelial monolayers in culture cups. Transfer cups from their culture wells to the EndOhm chamber for measurement rather than using hand-held electrodes. The chamber and the cap each contain a pair of concentric electrodes: a voltage-sensing silver/silver chloride pellet in the center plus an annular current electrode.
Catalog No. NC0590528