Learn More
Discovery Life Sciences Endometrial Ca Active Tx PBMC
SDP
Supplier:  Discovery Life Sciences 121395
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04037