Learn More
CPC Scientific H-Tyr-Pro-Phe-Phe-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ENDM002B
Encompass
Peptide that, together with endomorphin-1, has the highest affinity and specificity for the mu-opiate receptor and potent mu-selective bioactivity, including analgesia.ONE-LETTER SEQUENCE: YPFF-NH2MOLECULAR FORMULA: C32H37N5O5MOLECULAR WEIGHT:571.68STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [141801-26-5], RESEARCH AREA: PainREFERENCES: J. Zadina et al, Nature, 386, 499, (1997)
Catalog No. 50-210-9224