Learn More
Cygnus Technologies EndonucleaseGTP Antigen Concentrate, Essential For Quality Control, Variable Volume
Supplier:  Cygnus Technologies F963X
Encompass
The Cygnus Technologies EndonucleaseGTP Antigen Concentrate is a vital reagent for biopharmaceutical applications. It aids researchers in validating assay performance and accurately detecting residual endonuclease impurities in viral vector and vaccine preparations. This concentrate serves as a positive control for spike and recovery studies, ensuring reliable results in assays.
Catalog No. NC2176348