Learn More
Medchemexpress LLC Endoplasmic reticulum dye 1 | 1404104-40-0 | 99.8% | C21H12N7O3Re | 10MG
SDP
Supplier:  Medchemexpress LLC HY13621310MG
Encompass_Preferred
Endoplasmic reticulum dye 1 is a rhenium-based fluorescent probe for live-cell imaging of the endoplasmic reticulum and for detection of exocytotic events. It provides bright emission with low photobleaching and low cytotoxicity, and is supplied as a solid or as a DMSO solution for fluorescence microscopy applications.
  • Bright fluorescence with excitation at 463 nm and emission at 498 nm.
  • High purity (99.8%).
  • Low photobleaching for extended imaging sessions.
  • Low cytotoxicity suitable for live-cell experiments.
  • Soluble in DMSO at high concentration for easy preparation.
  • Available as solid or pre-made DMSO solution for convenience.
Catalog No. 50-002-61859