Learn More
Worthington Biochemical Corporation ENDOPROTEINASE GLU-C 1MG
SDP
Supplier:  Worthington Biochemical Corporation LS003608
Encompass
Protease, S. Aureus (Endoproteinase Glu-C), Code: STAP, Minimum activity: 500U/mg dry weight, A lyophilized powder, Store at 2 deg. to 8 deg.C, 1mg
Catalog No. NC9346805