Learn More
Medchemexpress LLC Endoproteinase Lys-C | 72561-05-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP3208A5MG
Encompass_Preferred
Endoproteinase Lys-C is a protease that cleaves proteins on the C-terminal side of lysine residues and is commonly used for protein sequencing. It is a key tool in biochemical research for understanding protein structure and function.
  • Cleaves proteins at lysine residues
  • Essential for protein sequencing applications
  • Supports in-depth protein structure analysis
Catalog No. 50-003-99243