Learn More
Medchemexpress LLC β-Endorphin, equine TFA | 97.1% | 3423.94 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1866A10MG
Encompass_Preferred
Beta-Endorphin, equine TFA, is an endogenous opioid peptide known for its analgesic properties. It binds with high affinity to both mu and delta opioid receptors, and studies have shown its ability to inhibit striatal dopamine release in vitro and influence acquisition and retention in rats in vivo.
  • Endogenous opioid peptide
  • Binds to mu and delta opioid receptors with high affinity
  • Possesses analgesic properties
  • Inhibits striatal dopamine release in vitro
  • Affects acquisition and retention in rats in vivo
  • Sequence: Tyr-Gly-Gly-Phe-Met-Ser-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-His-Lys-Lys-Gly-Gln
Catalog No. 50-004-24327