Learn More
Med Vet International Endoscopy Cleaning Brush, 3mm dia x 100mm, J1636D9, Each
Supplier:  Med Vet International JOR1636D9
Encompass_Preferred
Endoscopy Cleaning Brush Cleaning Brush • For use in 2.0-2.6mm working channels Diameter 3mm Working Length 100mm
Catalog No. 50-005-83507