Learn More
Apexbio Technology LLC Endostatin (84-114)-NH2 (JKC367) 10mg
SDP
Supplier:  Apexbio Technology LLC A110910
Encompass_Preferred
Endostatin 84-114 -NH2 (JKC367) is a C-terminal fragment derived from collagen XVIII with angiogenesis-inhibiting activity It functions by suppressing endothelial cell proliferation and migration required for neovascularization Endostatin 84-114 -NH2 exerts its biological activity primarily through the inhibition of angiogenesis In experimental tumor models continuous local administration of Endostatin 84-114 -NH2 has been reported to potentiate antitumor activities at lower dosages compared to intermittent injection Based on these pharmacological properties Endostatin 84-114 -NH2 (JKC367) holds research potential in oncology for tumor growth regulation metastatic process investigation and in vascular remodeling studies in cardiovascular disease models such as atherosclerosis
Catalog No. 50-000-96710