Learn More
Medchemexpress LLC Endothelial lipase inhibitor-1 | 1466427-02-0 | 98.0% | 406.43 g/mol | C22H22N4O4 | 5 MG

Supplier: Medchemexpress LLC HY1129115MG
Endothelial lipase inhibitor-1 is a small-molecule research compound that potently inhibits endothelial lipase (IC50 49 nM). It is supplied for laboratory research use only and is provided in solid and DMSO solution formats, with analytical support documents available for quality verification.
- Potent endothelial lipase inhibition (IC50 49 nM).
- High purity (98.02%) suitable for research use.
- Available as solid and ready-to-use DMSO solution formats.
- Soluble in DMSO (25 mg/mL) and formulatable for in vivo dosing.
- Includes supporting documents: data sheet, COA, SDS, HNMR, LCMS.
- Recommended storage: powder -20 °C for long term; in solvent -80 °C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.