Learn More
Medchemexpress LLC Endothelial protein C receptor (EPCR) | >95.0% | 100 UG

Supplier: Medchemexpress LLC HYP75244100UG
Recombinant rat endothelial protein C receptor (EPCR) expressed in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized for research use, the protein is provided with purity and formulation data and recommended storage instructions for biochemical and receptor-binding assays.
- Expressed in HEK293 cells for mammalian post-translational modifications.
- Contains a C-terminal 6xHis tag to facilitate purification.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Lyophilized from 0.22 μm filtered PBS, pH 7.4, for stability during storage and transport.
- Sequence accession Q4V8I1, covering residues N21-S213.
- For research use only; not for human therapeutic or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.