Learn More
CPC Scientific H-Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH (ammonium acetate salt) 5MG
SDP
Supplier:  CPC Scientific ENDT001C
Encompass
Potent vasoconstrictor that acts on endothelia cells and on the underlying smooth muscle cells as a modulator of vascular tone. ET-1 displays selectivity for ETA receptor.ONE-LETTER SEQUENCE: CSCSSLMDKECVYFCHLDIIW (C1&C15 bridge, C3&C11 bridge)MOLECULAR FORMULA: C109H159N25O32S5MOLECULAR WEIGHT:2492STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117399-94-7], SYNONYMS: ET-1 (human, bovine, dog, mouse, porcine, rat), Endothelin-1 (human, bovine, dog, mouse, porcine, rat)RESEARCH AREA: CardiovascularREFERENCES: A. Inoue, et al., PNAS, 80, 2863 (1989); M. Yanagisawa et al., TIPS, 10, 374 (1989); G.J. Price et al., Nucleic Acids Res., 18, 3658 (1991)
Catalog No. 50-210-9237