Learn More
CPC Scientific H-Cys-Ser-Cys-Ser-Ser-Trp-Leu-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ENDT004A
Encompass
SEQUENCE: H-Cys-Ser-Cys-Ser-Ser-Trp-Leu-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH (trifluoroacetate salt)(Cys1 and 15 bridge, Cys3 and 11 bridge)ONE-LETTER SEQUENCE: CSCSSWLDKECVYFCHLDIIW (C1&C15 bridge, C3&C11 bridge)MOLECULAR FORMULA: C115H160N26O32S4MOLECULAR WEIGHT: 2546.94STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [123562-20-9]SYNONYMS: RESEARCH AREA: CardiovascularREFERENCES:A. Inoue, et al., PNAS, 86, 2863 (1989)SKU(s): ENDT-004A
Catalog No. 50-210-9238