Learn More
ILEX LIFE SCIENCES LLC EndoTrap HD 10, 10 mL settled resin, Affinity chromatography endotoxin (LPS) removal resin
SDP
Supplier:  ILEX LIFE SCIENCES LLC LET0011
Encompass_Preferred
EndoTrap HD endotoxin removal products, manufactured in Germany by LIONEX, are intended for in vitro quantitative removal of lipopolysaccharide (LPS) from biological samples in aqueous solutions such as proteins, antibodies, cell extracts, phage, and nucleic acids. EndoTrap HD can be used in small scale processes for R&D and large scale manufacturing processes. It can be applied in early or late biomanufacturing process steps. EndoTrap HD is based on hydrophilic, dimensionally stable affinity matrix with excellent pressure/flow characteristics.Method: affinity chromatography.Format: pre-filled gravity flow columns, pre-filled FPLC columns, or bulk resin.LPS Binding Capacity: > 5,000,000 EU/mL.LPS Removal Rate: > 99.99%.Sample Recovery Rate: > 95%.pH Sample Range: 4-10.Non-toxic, no cytotoxic effects.Re-use more than 10 times.Suitable for high-salt conditions.The EndoTrap HD Leakage ELISA is available for quantitative detection of leached ligand from EndoTrap HD resin.
Catalog No. 50-212-0017