Learn More
BioVendor, LLC EndoTrap HD 5ml FPLC Column for endotoxin removal
SDP
Supplier:  BioVendor, LLC LET0067
Encompass
EndoTrap HD 5ml FPLC Column for endotoxin removal. EndoTrap HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples. The EndoTrap HD resin is designed to meet the needs of pharma and biotech industry for the efficient removal of endotoxins in biomanufacturing processes and any application that requires high endotoxin removal rates. EndoTrap HD fulfills all requirements for industrial applications, and can easily be integrated in early or late-stage purification processes. It is also available for small-scale purification as pre-filled columns. Manufactured by LIONEX GmbH in Germany. FEATURES: Best-in-class endotoxin binding capacity of >5,000,000 EU/ml. Removes >99.99% of LPS. Excellent sample recovery rate >95%. Non-toxic bacteriophage-derived ligand, no cytotoxic effects. Re-use >10 times. pH sample range: 4-10. Suitable for high salt conditions. Suitable for TRIS, HEPES, MOPS or similar buffered samples.
Catalog No. 50-196-3847