Learn More
Bachem Endotrophin (mouse) Trifluoroacetate
SDP
Supplier:  Bachem 4085553.0500
Encompass_Preferred
Endotrophin (mouse) Trifluoroacetate, H-Thr-Glu-Pro-Leu-Phe-Leu-Thr-Lys-Thr-Asp-Ile-Cys-Lys-Leu-Ser-Arg-Asp-Ala-Gly-Thr-Cys-Val-Asp-Phe-Lys-Leu-Leu-Trp-His-Tyr-Asp-Leu-Glu-Ser-Lys-Ser-Cys-Lys-Arg-Phe-Trp-Tyr-Gly-Gly-Cys-Gly-Gly-Asn-Glu-Asn-Arg-Phe-His-Ser-Gln-Glu-Glu-Cys-Glu-Lys-Met-Cys-Ser-Pro-Glu-Leu-Thr-Val-OH, 1678414-54-4, C345H520N92O106S7, Mr 7876.94, 0.5mg. Endotrophin is a carboxyl-terminal cleavage product of collagen 6 alpha-3 chain which is produced and released by adipocytes and massively upregulated in malignant tumors of breast, liver colon and pancreas. Endotrophin provides a link between obesity and aggressive tumor growth and may serve as sensitive diagnostic marker and valid therapeutic target for designing better strategies to treat cancer and ameliorate obesity-induced insulin resistance.
Catalog No. 50-194-6734