Learn More
Medchemexpress LLC Endoxifen-d5 | 1185244-45-4 | 97.5% | 378.52 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY18719ES1MG
Encompass_Preferred
Endoxifen-d5 is the deuterium labeled Endoxifen. Endoxifen is a key active metabolite of tamoxifen (TAM) with higher affinity and specificity to estrogen receptor that also inhibits aromatase activity. Endoxifen has the potential for breast cancer study.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88231