Learn More
Selleck Chemical LLC Endoxifen E-isomer S0712-25MG
SDP
Supplier:  Selleck Chemical LLC S071225MG
Encompass_Preferred
Endoxifen E-isomer (E-Endoxifen) an E-isomer of Endoxifen and impurity in Endoxifen Z-isomer drug substance exhibits antiestrogenic effects
Catalog No. 50-000-41796