Learn More
Medchemexpress LLC HY-18719D 5mg Medchemexpress, Endoxifen (E-isomer) CAS:114828-90-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18719D5MG
Encompass_Preferred
Medchemexpress, HY-18719D 5mg Endoxifen (E-isomer) CAS:114828-90-9 Endoxifen E-isomer is the E-isomer of (Z)-Endoxifen. (Z)-Endoxifen, an active metabolite generated via actions of CYP3A4/5 and CYP2D6, is a more potent selective estrogen receptor modulator (SERM) than Tamoxifen. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0011