Learn More
Medchemexpress LLC Endoxifen (hydrochloride) | 1197194-41-4 | 98.0% | 409.95 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY18719B100MG
Encompass_Preferred
Endoxifen hydrochloride is a key active metabolite of Tamoxifen with higher affinity and specificity to estrogen receptor that also inhibits aromatase activity. It has potential for breast cancer study.
  • More potent ER antagonist than tamoxifen.
  • Induces ER-alpha degradation and inhibits ER transcriptional activity.
  • Targets estrogen receptor α for degradation in breast cancer cells, inhibiting proliferation.
  • Exhibits strong growth inhibitory and cytotoxic effects on breast cancer cell lines at 10 μM.
  • Rapidly absorbed and systemically available; leads to higher exposure levels than tamoxifen.
  • Safe and effective in inhibiting human mammary tumor xenografts in female mice orally.
Catalog No. 50-003-87867