Learn More
Medchemexpress LLC Endoxifen Z-isomer hydrochloride | 1032008-74-4 | 99.67% | 409.95 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY187195MG
Encompass_Preferred
Endoxifen Z-isomer hydrochloride is an orally active Tamoxifen metabolite that acts as a selective estrogen receptor modulator. It is significantly more potent than Tamoxifen, with 100-fold greater efficacy. This compound inhibits PKCβ1 kinase activity and demonstrates anticancer properties against hormone-resistant metastatic breast cancer.
  • Inhibits PKCβ1 kinase activity
  • Reduces AKTSer473 phosphorylation
  • Diminishes AKT substrate phosphorylation
  • Induces apoptosis
  • Potent antiproliferative activity against MCF7AC1 and MCF7LR cell lines
  • Shows better antitumor efficacy in MCF7LR resistant mouse model
Catalog No. 50-003-88204