Learn More
BioVendor, LLC ENDOZYME II GO STRIPS, fluorescence end-point assay, recombinant Factor C, pre-coated strips, assays for 1 to 21 samples
SDP
Supplier:  BioVendor, LLC 424107
Encompass
ENDOZYME II GO STRIPS, fluorescence end-point assay, recombinant Factor C, pre-coated strips, assays for 1 to 21 samples
Catalog No. NC3136991
Only null left
Add to Cart
Request a Quote