Learn More
MARIAN MEDICAL INC ENFit Bottle Adapter, Press In, 16.5-18mm, 50 Per Box
Supplier:  MARIAN MEDICAL INC BA08L
Encompass_Preferred
Bottle Adapter. ENFit Bottle Adapter. Marian Medical Inc. BA Series. Press in ENFit bottle adapter for medication bottle access. Available in stepped universal and single size configurations. ENFit connection supports enteral medication workflows. For use with medication bottles in clinical and home care settings. 50 per box.
Catalog No. NC3958245