Learn More
Medchemexpress LLC Enflicoxib | 251442-94-1 | 99.9% | 405.34 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19384
Encompass_Preferred
Enflicoxib (E 6087) is a nonsteroidal anti-inflammatory compound that selectively inhibits cyclooxygenase-2 (COX-2) but not cyclooxygenase-1 (COX-1). It demonstrates anti-inflammatory, analgesic, and antipyretic activities in animal models. Enflicoxib is the primary active compound, with a long elimination half-life and a relatively large volume of distribution. It is suitable for laboratory research.
  • Selectively inhibits cyclooxygenase-2 (COX-2)
  • Does not inhibit cyclooxygenase-1 (COX-1)
  • Exhibits anti-inflammatory properties
  • Provides analgesic and antipyretic activities in animal models
  • Has a long elimination half-life
Catalog No. 50-003-85071