Learn More
Medchemexpress LLC Enfortumab vedotin-ejfv (solution) | 1346452-25-2 | 152,000 (average) | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP99016B1MG
Encompass_Preferred
Enfortumab vedotin-ejfv is an antibody-drug conjugate (ADC) for research use that targets Nectin-4. It consists of a humanized IgG1 antibody linked to the cytotoxic agent monomethyl auristatin E (MMAE) via a protease-cleavable valine-citrulline linker and is provided as a solution in a 1 mg pack for laboratory studies.
  • Targets nectin-4 with a humanized IgG1 antibody.
  • Conjugated to MMAE via a valine-citrulline (Val-Cit) protease-cleavable linker.
  • Provided as a solution suitable for in vitro research applications.
  • Supplied in a 1 mg pack size for laboratory experiments.
  • Includes a product data sheet with handling and composition details.
Catalog No. 50-002-03508