Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Supplier:  Grainger 16R995
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R966 is a carbide engraving tool with a single cutting end. It has a shank diameter of 1/8" and an overall length of 2". The carbide cutter features a 30 degree included angle. This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detail engraving work and making small letters or numbers. The Micro 100 engraving tool comes with a TiAIN coating for increased tool life and lubricity. It is made in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-3717