Learn More
Grainger Engraving Tool 10.00mm L of Cut Carbide
Supplier:  Grainger 16T023
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16T023 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". The tool is made of solid carbide for wear resistance when engraving into materials like steel, aluminum, and plastics. It can be used for detail engraving work in machines like CNC mills and lathes. The single end cutting edge allows for engraving by interpolating the workpiece past the rotating tool.
Catalog No. 50-276-3961