Learn More
Grainger Engraving Tool 10.00mm L of Cut Carbide
Supplier:  Grainger 16R958
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R958 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point for detail engraving in materials like plastics, wood, fiberglass and soft metals. It can be used in low horsepower, high speed engraving machines. The tool has a Rockwell hardness of C64-66 for wear resistance during engraving.
Catalog No. 50-276-4580