Learn More
Grainger Engraving Tool 10.00mm L of Cut Carbide
Supplier:  Grainger 16R960
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R960 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 3", and a cutting diameter of 1/16". This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can be used in low-speed handheld rotary tools or flex shaft machines. The single end cutting edge allows for detail work in tight areas. Made in the USA from M2 high-speed steel for good wear resistance and edge retention.
Catalog No. 50-276-3876