Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Supplier:  Grainger 16T036
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16T036 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is made of solid carbide for wear resistance and has a TiAIN coating for heat resistance and lubricity. It can machine a variety of materials including steel, aluminum, brass, copper, plastics, and wood. The single end cutting edge allows for detail work in corners and confined areas. This is a high quality carbide engraving tool well-suited for detailed engraving, routing, and milling applications.
Catalog No. 50-276-4013