Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Supplier:  Grainger 16R970
Encompass_Preferred
The Micro 100 Engraving Tool is a 12mm carbide engraving cutter with a 0.5mm corner radius. It has a right hand spiral flute and is designed for engraving, routing and profiling in materials like plastics, wood, fiberglass and soft metals. The tool has a 1/4 inch shank diameter and is made of solid carbide for wear resistance and long tool life. It can be run at speeds up to 45,000 rpm with light to medium chip loads. The overall length of the cutter is 1.5 inches.
Catalog No. 50-276-4633