Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Supplier:  Grainger 16T033
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16T033 is a carbide engraving tool with a single cutting end. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". The tool is made of solid carbide for wear resistance when engraving into materials like steel, aluminum, and plastics. It can be used in CNC machines or manual engraving tools. The single cutting end allows for detail work in tight areas. This is a high quality carbide engraving tool made in the USA by Micro 100.
Catalog No. 50-276-3906