Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Supplier:  Grainger 16R860
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R863 is a high-speed steel engraving cutter with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detail work, lettering, marking and engraving applications. The single end cutting edge allows for engraving in corners and confined areas. Made in the USA from M2 high speed steel for good wear resistance and heat tolerance.
Catalog No. 50-276-6745