Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Supplier:  Grainger 16R863
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R863 is a carbide tipped engraving tool designed for hand engraving, die sinking, and cavity sinking applications in most ferrous and non-ferrous materials. It has a single cut carbide tip brazed to a 3/16" diameter high speed steel shank. The cut is 1/32" across. It can be used at speeds up to 15,000 RPM with light to medium pressure. This is a high quality precision engraving tool made in the USA by Micro 100.
Catalog No. 50-276-7447