Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Supplier:  Grainger 16R983
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R966 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 30 degree included angle and is designed for hand engraving in materials like plastics, soft metals, wood and fiberglass. It can be used in low-powered rotary hand engraving machines. The tool has a Rockwell hardness of C60-C65 for wear resistance. It allows for fine detail engraving work.
Catalog No. 50-276-4094