Learn More
Grainger Engraving Tool 5.00mm L of Cut Carbide
Supplier:  Grainger 16R891
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R891 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 4 inches. The tool features a 135 degree point angle for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM. The tool has a bright finish for visibility and is made in the USA by Micro 100.
Catalog No. 50-276-4048