Learn More
Grainger Engraving Tool 5.00mm L of Cut Carbide
Supplier:  Grainger 16R889
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R889 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". The tool is made of solid carbide for wear resistance and has a right hand spiral flute. It is designed for light duty engraving in materials like plastics, wood, and soft metals. The single end cutting edge allows for engraving in tight spaces. This Micro 100 engraving tool is suitable for detailed engraving work where precision is required.
Catalog No. 50-276-4576