Learn More
Grainger Engraving Tool 5/8 L of Cut Carbide
Supplier:  Grainger 16T012
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16T012 is a double-ended carbide engraving tool made in the USA by Micro 100. It has a 1/8" shank diameter and comes in a set of 2, with one tool having a 30 degree included angle and the other having a 60 degree included angle. The overall tool length is 2-1/2". This versatile tool can be used for detail engraving work in most machinable materials.
Catalog No. 50-276-3957