Learn More
Grainger Engraving Tool 6.00mm L of Cut Carbide
Supplier:  Grainger 16R895
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R895 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point angle and is designed for hand engraving in materials like plastics, soft metals, wood and fiberglass. It can be used in low-speed handheld rotary tools or flex shaft machines up to 15,000 RPM. The tool's high-speed steel construction provides good wear resistance.
Catalog No. 50-276-4046