Learn More
Grainger Engraving Tool 7.00mm L of Cut Carbide
Supplier:  Grainger 16T020
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16T020 is a double-ended carbide engraving tool designed for engraving, scribing, and deburring in CNC machining applications. It features a 16 degree included angle and 0.020" tip diameter. The overall length is 3" and the shank diameter is 1/8". This double-ended tool provides two cutting edges for extended tool life. It is made of solid carbide for wear resistance when engraving in steel, aluminum, and other materials. The 16 degree tip angle is optimized for fine detail engraving work.
Catalog No. 50-276-3985