Learn More
Grainger Engraving Tool 7/16 L of Cut Carbide
Supplier:  Grainger 16R922
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R922 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 4 inches. The tool features a 135 degree point for detail engraving in materials like plastics, wood, fiberglass and soft metals. It can be used in low horsepower engraving machines and rotary hand engravers. The tool's precision ground cutter provides a sharp cutting edge and smooth finish.
Catalog No. 50-276-3934