Learn More
Grainger Engraving Tool 8.00mm L of Cut Carbide
Supplier:  Grainger 16R904
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16R904 is a double-ended carbide engraving tool made in the USA by Micro 100. It has a 1/8" shank diameter and comes in a set of 2, with one end having a 30 degree included angle and the other end having a 60 degree included angle. The overall tool length is 2-1/2". This versatile tool can be used for detail engraving work in most machinable materials.
Catalog No. 50-276-4573