Learn More
AdipoGen EnhancedFasL Soluble h Pack
SDP
Supplier:  AdipoGen AG44B0001KI01
Encompass_Preferred
Set. FasL, Soluble human rec. induces apoptosis in a concentration range of <1ng/ml in the presence of 0. 1 to 1µg/ml of TNF Ligands Enhancer Prod. No. AG-35B-0001. Optimal conditions must be determined individually for each cell line tested
Catalog No. 50-256-2754