Learn More
Anton Paar Enjoy a kinematic viscometer with an intuitive interface and compact design. Measure from 15–100 °C and get digital results in under 5 minutes using just 1.5 mL—no stopwatch or manual calculations needed
Supplier:  Anton Paar 105010
Encompass_Preferred
Test Methods: ASTM D7042, D445 bias corrected, ISO 23581, EN 16896Measuring Range:Viscosity: 0.3 mm²/s to 5,000 mm²/sTemperature: +15 °C to +100 °C (one temperature, second temperature optional)Viscosity repeatability** and reproducibility**0.1 % and 0.35 %Temperature repeatability: 0.005 °CTemperature reproducibility: 0.03 °C from +15 °C to +100 °CSample volume min. / typical: 1.5 mL / 5 mLSolvent volume min. / typical: 1.5 mL / 6 mLMax. sample throughput: 37 samples per hourOptional upgrades: Second temperatureAccessories: Magnetic particle trap (MPT)
Catalog No. 50-005-80465
Request a Quote
Please contact your sales representative for a quote or further ordering instructions.