Learn More
Medchemexpress LLC Enlicitide (chloride) | 2407527-16-4 | 99.6% | 1586.29 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP4153500UG
Encompass_Preferred
Enlicitide chloride is an orally active PCSK9 inhibitor with an IC50 of 2.5 nM. It blocks the interaction between the LPL receptor and PCSK9, increasing LPL receptor levels and ultimately decreasing low-density lipoprotein cholesterol (LDL-C) levels. It is used in the study of cardiovascular diseases such as atherosclerosis and hypercholesterolemia.
  • Orally active PCSK9 inhibitor
  • Blocks interaction between LPL receptor and PCSK9
  • Increases LPL receptor levels
  • Decreases low-density lipoprotein cholesterol (LDL-C) levels
  • Suitable for cardiovascular disease research
Catalog No. 50-003-97265