Learn More
Medchemexpress LLC Enmein | 3776-39-4 | 1 MG

Supplier: Medchemexpress LLC HYN50281MG
Enmein, isolated from *I. serra*, is a chemical compound with immunosuppressive effects. It has a molecular formula of C20H26O6 and a molecular weight of 362.42. This white to off-white solid is intended for research use only.
- Purity of 98.0%
- Terpenoid, diterpenoid classification
- Storage at -20°C, protected from light
- Soluble in DMSO
- Exhibits antiproliferative and cytotoxic activity against various human cancer cell lines
- Immunosuppressive effect on lymphocytes and in mouse models
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.