Learn More
Cayman Chemical Ennatn B1 5mg
SDP
Supplier:  Cayman Chemical 172455 MG
Encompass_Preferred
A cyclohexadepsipeptide that has been shown to induce apoptosis in several cancer lines (EC50s ≤ 10 µM) and to decrease the activation of the cell proliferation kinase, ERK (p44/p42); inhibits the multi-drug resistance transporter Pdr5p from S. cerevisiae
Catalog No. 50-297-9595